Express Eingabe
Austria
AuftragsverfolgungKontakt
Split/splitless inlet seals
Optional split/splitless weldment
  HEWLG1544-60585
 :   Agilent
Split/splitless inlet seals
  HEWLG1544-60585
 :  Agilent
Split/splitless inlet seals
Split/splitless inlet seals
Optional split/splitless weldment
817,00 €
1 * 1 ST
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
  • Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
  • High quality, leak-free seal
 : An inlet washer is provided with each inlet seal.
  • Bezeichnung:
    Optional split/splitless weldment
  • Seq:
    10
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.