Express Eingabe
Austria
AuftragsverfolgungKontakt
AccuGENE® 1M Tris HCl buffer | Avantor

AccuGENE® 1M Tris HCl buffer

AccuGENE™ 1 M Tris HCl buffer, 7,2, 1 L
Artikelnr. 733-1652
Anbieter:   LONZA
AccuGENE® 1M Tris HCl buffer
Artikelnr. 733-1652
Anbieter:  LONZA
CAS Nummer:  
AccuGENE® 1M Tris HCl buffer

AccuGENE™ 1 M Tris HCl buffer, 7,2, 1 L

CAS Nummer:  
Preis
110,00 €
Einloggen für Verfügbarkeit
1 * 1 L
Product Details & Documents

AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.

  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs

Tris-HCl is a solution containing tris base mixed with hydrocholoric acid. Tris has a pKa of 8,07 at STP and is used in bufferings solutions to prevent large changes in pH level. A staple of electrophoretic and other biological applications, Tris has a useful buffer range of 7 to 9, which coincides with the physiological pH typical of most living organisms. Tris is the 'T' in TAE and TBE Buffers.

Each reagent is prepared with 18 megOhm water, filtered using a 0,2 micron filter, and filled into sterile bottles.

Store all reagents at 18 to 26°C

Dokumente

Spezifikationen

Spezifikationen

  • VE:
    1 L
  • Bezeichnung:
    AccuGENE™ 1 M Tris HCl buffer
  • pH-Wert:
    7,2

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.