Spherical ground joints, Quickfit®
S 29, -
Artikelnr. 201-8205
Anbieter: SciLabware
Springe zu:
Product Details & Documents
Made of PYREX®, borosilicate glass.
- Excellent sealing properties
- Minimal requirement for joint lubrication
- Ideal for use as flexible connectors
For the Glassblower who demands the highest quality joint.
Length: 100 mm
Spezifikationen
- Kugelschliff-Kugel:S 29
- Kugelschliff-Schale:-