Spherical ground joints, Quickfit®
S 19, -
Artikelnr. 201-8204
Anbieter: SciLabware
Springe zu:
Product Details & Documents
Made of PYREX®, borosilicate glass.
- Excellent sealing properties
- Minimal requirement for joint lubrication
- Ideal for use as flexible connectors
For the Glassblower who demands the highest quality joint.
Length: 100 mm
Spezifikationen
- Kugelschliff-Kugel:S 19
- Kugelschliff-Schale:-
- Cat. No.:201-8204