Pulverkolben
2000 ml, 29/32 NS,
Artikelnr. LENZ03043073
Anbieter: Lenz Laborglas
Springe zu:
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for rotary evaporators, for drying of powder-like substances. With center neck, conical joint and inscription label.
Dokumente
BSE TSE Glass Columns
BSE/TSE-Zertifikate
Spezifikationen
- Hülse:29/32 NS
- Kapazität:2000 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.