Doppelspatel, breit
300 mm, 30 mm
Artikelnr. RSGA092.300
Anbieter: RSG
Springe zu:
Product Details & Documents
Double spatulas with wide, flat endings. Made of stainless steel.
Documents
RSGA092.500_TDS_EN
Datasheet
Spezifikationen
- Länge:300 mm
- Klingenbreite:30 mm