Chemikalienlöffel
Chemikalienlöffel, zylindrischer Stiel, runder Löffel, 150 mm
Artikelnr. RSGA024.150
Anbieter: RSG
Springe zu:
Product Details & Documents
Single-ended chemical spoons made of stainless steel.
- Round handle
Spezifikationen
- Bezeichnung:Chemikalienlöffel, zylindrischer Stiel, runder Löffel
- Länge:150 mm
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.