AuftragsverfolgungKontakt
Forceps
Forceps
Forceps for insect pins, with ratchet, bent, square tips, 155 mm
Catalog # HAMMHSC030-15
Supplier:   HAMMACHER KARL
Forceps
Catalog # HAMMHSC030-15
Supplier:  HAMMACHER KARL
Forceps
Forceps
Forceps for insect pins, with ratchet, bent, square tips, 155 mm
Price
79,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
Specifications
  • Typ:
    Forceps for insect pins, with ratchet, bent, square tips
  • Länge:
    155 mm
  • Seq:
    06