Express Eingabe
Austria
AuftragsverfolgungKontakt
Glass wool
Glass wool
Quartz wool, very fine fibers
Catalog # MANA718587
Supplier:   MACHEREY-NAGEL
Glass wool
Catalog # MANA718587
Supplier:  MACHEREY-NAGEL
Glass wool
Glass wool
Quartz wool, very fine fibers
Price
66,30 €
Log in for availability
1 * 25 g
Product Details & Documents
This glass wool can be used for GC liners, packed GC columns etc.
Specifications
  • Bezeichnung:
    Quartz wool, very fine fibers
  • Seq:
    03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.