Express Eingabe
Austria
AuftragsverfolgungKontakt
Glass wool
Glass wool, long fibers, DMCS treated, for packed GC columns
  MANA706201
Glass wool
  MANA706201
Glass wool
Glass wool
Glass wool, long fibers, DMCS treated, for packed GC columns
105,00 €
1 * 50 g
This glass wool can be used for GC liners, packed GC columns etc.
  • Bezeichnung:
    Glass wool, long fibers, DMCS treated, for packed GC columns
  • Seq:
    01