Extraction thimbles
68×250 mm, 2,0 mm
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
- Fat determination in food and feed
- Determination of binder content in asphalt
- Analysis of soil contaminants
- Determination of additives in rubber and elastomers
Specifications
- I-Ø×H:68×250 mm
- Seq:41
- Stärke:2,0 mm