AuftragsverfolgungKontakt
Extraction thimbles
15×100 mm, 1,0 mm
Catalog # MANA645004
Supplier:   MACHEREY-NAGEL
Extraction thimbles
Catalog # MANA645004
Supplier:  MACHEREY-NAGEL
Extraction thimbles
Extraction thimbles
15×100 mm, 1,0 mm
Price
97,70 €
Log in for availability
1 * 25 ST
Product Details & Documents
Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Specifications
  • I-Ø×H:
    15×100 mm
  • Seq:
    04
  • Stärke:
    1,0 mm