Tricin
5 kg
Artikelnr. A1085.5000
Anbieter: PanReac AppliChem
Springe zu:
Product Details & Documents
Synonyme: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
- CAS Nummer:5704-04-1
- Formel:(HOCH₂)₃CNHCH₂CO₂H
- MDL-Nummer:MFCD00004277
- Molekulargewicht:179.17 g/mol
- Merck-Index:13,09724
- Schmelzpunkt:187 °C
- Dichte:1,523 g/cm³ (19.85 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:227-193-6
Spezifikationen
Spezifikationen
- VE:5 kg
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



