Express Eingabe
Austria
AuftragsverfolgungKontakt
γ-(N-Morpholino)propansulfonsäure (MOPS)

γ-(N-Morpholino)propansulfonsäure (MOPS)

Glass bottle for solids, 500 g
Catalog # A2947.0500
Supplier:   PanReac AppliChem
γ-(N-Morpholino)propansulfonsäure (MOPS)
Catalog # A2947.0500
Supplier:  PanReac AppliChem
CAS Number:  1132-61-2
γ-(N-Morpholino)propansulfonsäure (MOPS)

Glass bottle for solids, 500 g

CAS Number:  1132-61-2
Price
382,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
  • CAS Number:1132-61-2
  • Formula:C₇H₁₅NO₄S
  • MDL Number:MFCD00006183
  • Molecular Weight:209.27 g/mol
  • Merck Index:13,06288
  • Melting Pt:> 250 °C
  • Flash Pt:110 °C (geschlossener Tiegel)
  • Storage Temperature:Raumtemperatur
  • EINECS:214-478-5

Documents

Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.