Express Eingabe
Austria
AuftragsverfolgungKontakt
Connectors, PP | Avantor
Connectors, PP
Connector, straight, female luer lock/tubing nipple, 4,8 mm
Artikelnr. INFIPP-LFS48
Anbieter:   PIEPER FILTER
Connectors, PP
Artikelnr. INFIPP-LFS48
Anbieter:  PIEPER FILTER
Connectors, PP
Connectors, PP
Connector, straight, female luer lock/tubing nipple, 4,8 mm
Preis
28,50 €
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents
Connectors, made of PP, natural.
  • Good chemical resistance
  • Good temperature resistance
Spezifikationen
  • Schlauch Innen-Ø:
    4,8 mm
  • Bezeichnung:
    Connector, straight, female luer lock/tubing nipple
  • Seq:
    11
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.