Deckgläser, rund
Round cover slides, 14 mm, Borosilikatglas
Artikelnr. 631-0692
Anbieter: MARIENFELD
Springe zu:
Product Details & Documents
Round cover glasses made of pure white borosilicate D 263 ™ M glass, Hydrolytic Class I.
- No bubbles or flaws
Suitable for in vitro diagnostic (IVD) applications.
: 100 cover glasses per box with separate lid.
Spezifikationen
- Ø:14 mm
- Bezeichnung:Round cover slides
- Material:Borosilikatglas
- Seq:004
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.