Trypsinising flasks, with screw caps, WHEATON®
150 ml, Schraubverschluss oben, 38-430
WHEA355444
Made of borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These trypsinising flasks units have screw caps with white styrene-butadiene rubber liners that reduce contamination of contents. Fluted design improves mixing.
: Conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
- Kapazität:150 ml
- Bezeichnung:Schraubverschluss oben, 38-430