Trypsinising flasks, with screw caps, WHEATON®
35 ml, Schraubverschluss oben, 24-430
Artikelnr. WHEA355442
Anbieter: DWK Life Sciences
Springe zu:
Product Details & Documents
Made of borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These trypsinising flasks units have screw caps with white styrene-butadiene rubber liners that reduce contamination of contents. Fluted design improves mixing.
Zertifizierung: Conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
Spezifikationen
- Kapazität:35 ml
- Bezeichnung:Schraubverschluss oben, 24-430
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.