ACM craft tools
Product Details & Documents
The ACM tool set is presented in a resilient blue carry case and contains a comprehensive range of 13 precision ground carbon steel blades manufactured to a high surgical quality.
Also including three handle variations the set can be used for general cutting applications including the trimming of light and medium weight materials, precision trimming, stencilling, scoring and etching as well as corner cuts, deburring, stripping and gasket cutting making this set extremely versatile.
ACM No. 1 handle - for light duty applications
A light duty craft knife designed to assist with smaller cutting projects on soft or lightweight material.
Suitable blades: No. 7 / 10 / 11 / 16 / 17
ACM No. 2 handle - for medium duty applications
Similar to the light weight knife but slightly thicker to provide added strength and stability.
Suitable blades: No. 2 / 18 / 22 / 24 / 28
ACM No. 5 handle - for heavy duty applications
A heavy duty craft knife that is larger and thicker than all the other types of knives.
Suitable blades: No. 2 / 18 / 22 / 24 / 28
Bestellinformation: All the blades and handles are available on an individual basis or as smaller sets.
Spezifikationen
- Für:Klingen Nr. 7/10/11/16/17
- Typ:Nr. 1
- Bezeichnung:ACM-Griff, mit Klinge Nr. 11, auf Blisterkarte
- Länge:146 mm
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.