AuftragsverfolgungKontakt
Threaded adapters, BOLA | Avantor
Threaded adapters, BOLA
Transition from GL40/S40 to GL 45
Artikelnr. BOHLH978-40
Anbieter:   Bohlender
Threaded adapters, BOLA
Artikelnr. BOHLH978-40
Anbieter:  Bohlender
Threaded adapters, BOLA
Threaded adapters, BOLA
Transition from GL40/S40 to GL 45
Preis
18,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Adaptors, made of PTFE, white, allow the use of BOLA multiple distributors for bottles with female thread GL 45 also on bottles with GL 32, GL 40 and S 40 threads.
  • Chemical resistant
  • Temperature resistant from −200 to +200 °C
Spezifikationen
  • Bezeichnung:
    Transition from GL40/S40 to GL 45
  • Seq:
    02