Hose clips, worm-threaded
8 - 12 mm
Artikelnr. BURK8677-0812
Anbieter: BURKLE
Springe zu:
Product Details & Documents
Hose clip sleeve made of stainless steel 1,4016, screw zinc coated, with screw thread.
Dokumente
BURK8677-0812_TDS_EN BURK8677-1016_TDS_EN BURK8677-1220_TDS_EN
Technisches Datenblatt
Spezifikationen
- Spannweite:8 - 12 mm
- Seq:01