Bone curettes, Aesculap®
Bone curette, type Daubenspeck (Fig. 0000), with 2,8 mm end, angled upwards, 200 mm
Artikelnr. AESCFK662R
Anbieter: Aesculap
Springe zu:
Product Details & Documents
Spezifikationen
- Bezeichnung:Bone curette, type Daubenspeck (Fig. 0000), with 2,8 mm end, angled upwards
- Länge:200 mm
- Seq:30
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



