Micro scissors for endoscopic work, Aesculap® MINOP®
Replacement inner tube for MINOP instrument FF386R
Artikelnr. AESCFF436R
Anbieter: Aesculap
Springe zu:
Product Details & Documents
Rigid micro scissors for advanced intraventricular Neuroendoscopy.
- Very delicate
Ø = 2 mm
Ordering information: Complete instrument consists of a handle, an outer tube, and a jaw part with inner tube. Replacements can be ordered separately.
Spezifikationen
- Länge:Replacement inner tube for MINOP instrument FF386R
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.