Micro scissors for endoscopic work, Aesculap® MINOP®
Replacement handle for MINOP instruments FF385R - FF389R
Artikelnr. AESCFF432R
Anbieter: Aesculap
Springe zu:
Product Details & Documents
Rigid micro scissors for advanced intraventricular Neuroendoscopy.
- Very delicate
Ø = 2 mm
Bestellinformation: Complete instrument consists of a handle, an outer tube, and a jaw part with inner tube. Replacements can be ordered separately.
Spezifikationen
- Länge:Replacement handle for MINOP instruments FF385R - FF389R
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.