Dissecting forceps, Aesculap®
Dissecting forceps, Angled, serrated, very fine tips, 200 mm
Catalog # AESCBD257R
Supplier: Aesculap
Jump to:
Product Details & Documents
Stainless steel.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
Delicate dissecting forceps with plateau and many types.
Specifications
- Typ:Angled, serrated, very fine tips
- Bezeichnung:Dissecting forceps
- Länge:200 mm
- Seq:16