Dissecting forceps, Aesculap®
Dissecting forceps, Cushing, Straight, serrated, very fine tips, 200 mm
Artikelnr. AESCBD239R
Anbieter: Aesculap
Springe zu:
Product Details & Documents
Stainless steel.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
Delicate dissecting forceps with plateau and many types.
Spezifikationen
- Typ:Straight, serrated, very fine tips
- Bezeichnung:Dissecting forceps, Cushing
- Länge:200 mm
- Seq:20