Express Eingabe
Austria
AuftragsverfolgungKontakt
Pots, laboratory, without lid
Lids for laboratory pots, stainless steel 18/10, for 3,5 l pots
Artikelnr. BOCH8371
Anbieter:   BOCHEM
Pots, laboratory, without lid
Artikelnr. BOCH8371
Anbieter:  BOCHEM
Pots, laboratory, without lid
Pots, laboratory, without lid
Lids for laboratory pots, stainless steel 18/10, for 3,5 l pots
Preis
32,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Stainless steel 18/10.
  • Rounded corners for fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance
Multipurpose pots for a wide range of uses. This series is suitable for induction fields.
 : Lids to be ordered separately.
Spezifikationen
  • Bezeichnung:
    Lids for laboratory pots, stainless steel 18/10, for 3,5 l pots
  • Seq:
    10
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.