Reading pocket magnifiers
Einschlaglupe in schwarzem Lederetui, 3,5×, 60 mm
Artikelnr. ESCH1752560
Anbieter: ESCHENBACH OPTIK
Springe zu:
Product Details & Documents
These handheld folding magnifiers with classic design fit easily into any handbag or pocket.
- Many different colours and sizes
- Anti-scratch coating for both models
- Black or brown leather case available
Spezifikationen
- Ø:60 mm
- Vergrößerung:3,5×
- Bezeichnung:Einschlaglupe in schwarzem Lederetui