Amidoschwefelsäure ≥99%, TECHNICAL
Kunststoffeimer, 5 kg
: Aminosulphonic acid, Amidosulphuric acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid
- :5329-14-6
- :H₃NO₃S
- :MFCD00011603
- :97.09 g/mol
- :13,09006
- :247 °C (1013 hPa)
- :205 °C
- :2,15 g/cm³ (20 °C)
- :Raumtemperatur
- :226-218-8
- :2967
- :8,III
Documents
1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 30L Plastic bucket UN Z (ZPACK13247033) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
P20_VWRC20671.297_SPEC_DE.pdf P20_VWRC20671.366_SPEC_DE.pdf spec_VWRC20671.297 spec_VWRC20671.297_de spec_VWRC20671.366
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
- Assay:Min. 99 %
- Packung:Kunststoffeimer
- VE:5 kg