Express Eingabe
Austria
AuftragsverfolgungKontakt
Eigelbemulsion

Eigelbemulsion

Eigelbemulsion, 50%, 4,5 L
Catalog # EY4500
Supplier:   BALIS
Eigelbemulsion
Catalog # EY4500
Supplier:  BALIS
CAS Number:  
Eigelbemulsion
Eigelbemulsion

Eigelbemulsion, 50%, 4,5 L

CAS Number:  
Price
654,00 €
Log in for availability
1 * 4,5 L
Product Details & Documents

Zusatzstoff zu Cereus-Selektivagarbasis nach Mossel.

  • Bestrahlt

Documents

Specifications

Specifications

  • VE:
    4,5 L
  • Bezeichnung:
    Eigelbemulsion, 50%

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.