Büchner-Trichter
120 mm, 1000 ml, 3
511-0780
PYREX®, borosilicate glass, with sintered glass disc.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Very good temperature resistance
For convenient filtration of solutions.
- Platte Ø:120 mm
- Porosität:3
- Kapazität:1000 ml
- Seq:49



