Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Microplate stands

Well plate stand, tilted at 30° angle
Artikelnr. DIVEWPST-1000
Anbieter:   DIVERSIFIED BIOTECH
Microplate stands
Artikelnr. DIVEWPST-1000
Microplate stands

Well plate stand, tilted at 30° angle

Preis
136,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Convenient holder for microplates to comfortably view samples and pipette without strain. Available tilted at a 30° angle or as an adjustable version allowing the flexibility to adjust the angle of the microplate (45° max. tilt).

Spezifikationen
  • Bezeichnung:
    Well plate stand, tilted at 30° angle
  • Seq:
    02