Combs for gel casting system, MultiCaster
Multicaster comb, orange
Catalog # 730-0363
Supplier: EMBITEC
Jump to:
Product Details & Documents
Reversible combs for the MultiCaster gel casting system with dual configuration for multiple gel formats.
- Made of polycarbonate
- Well configurations and estimated sample volume: 12+1 well (30 μl) and 24+1 well (11 μl)
- Multi-channel pipette compatibility: 12-channel
- Estimated sample volume is based on gel thickness of 6,5 mm
All combs are interchangeable so that a single MultiCaster system can be used for all preparations and high throughput needs by simply swapping the combs.
Specifications
- Bezeichnung:Multicaster comb, orange
- Seq:03