Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Measuring Cylinders

10 ml, 0,2 ml, 140 mm, 0,2 ± ml
  612-0426
 :   SciLabware
Measuring Cylinders
  612-0426
Measuring Cylinders
Measuring Cylinders

10 ml, 0,2 ml, 140 mm, 0,2 ± ml

 
6,70 €
   
1 * 1 ST
 

PMP (TPX®), transparent, tall form, autoclavable, class B.

  • Hexagonal base with non-slip, anti-suction feature, spout, excellent chemical resistance, no meniscus to confuse readings
  • Recycling codes and batch code on each cylinder
  • Autoclavable - Does not affect accuracy

Note: The product is suitable for lab washing machines, however, a temperature of below 60°C should be used to prevent damage.

 : ISO 6706

 
  • Kapazität:
    10 ml
  • Höhe:
    140 mm
  • Teilung:
    0,2 ml
  • Toleranz:
    0,2 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.