Express Eingabe
Austria
AuftragsverfolgungKontakt
Büchner-Trichter

Büchner-Trichter

175 mm, 4000 ml, 1
Artikelnr. 511-0781
Anbieter:   SciLabware
Büchner-Trichter
Artikelnr. 511-0781
Anbieter:  SciLabware
Büchner-Trichter
Büchner-Trichter

175 mm, 4000 ml, 1

Preis
597,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

PYREX®, borosilicate glass, with sintered glass disc.

  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Very good temperature resistance

For convenient filtration of solutions.

Spezifikationen
  • Platte Ø:
    175 mm
  • Porosität:
    1
  • Kapazität:
    4000 ml
  • Seq:
    53
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.