Express Eingabe
Austria
AuftragsverfolgungKontakt
Ammoniumnitrat ≥98,5%, TECHNICAL

Ammoniumnitrat ≥98,5%, TECHNICAL

Kunststoffeimer, 5 kg
Catalog # 21277.361
Oxidizing
Supplier:   VWR Chemicals
Ammoniumnitrat ≥98,5%, TECHNICAL
Catalog # 21277.361
Supplier:  VWR Chemicals
CAS Number:  6484-52-2
Ammoniumnitrat ≥98,5%, TECHNICAL
Ammoniumnitrat ≥98,5%, TECHNICAL

Kunststoffeimer, 5 kg

CAS Number:  6484-52-2
Price
122,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
  • CAS Number:6484-52-2
  • Formula:NH₄NO₃
  • MDL Number:MFCD00011425
  • Molecular Weight:80.04 g/mol
  • Merck Index:13,00538
  • Boiling Pt:230 °C (1013 hPa)
  • Melting Pt:169 °C
  • Density:1,73 g/cm³ (20 °C)
  • Storage Temperature:Raumtemperatur
  • EINECS:229-347-8
  • UN:1942
  • ADR:5.1,III
Specifications

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 98.5 %

Specifications

  • Packung:
    Kunststoffeimer
  • VE:
    5 kg

Specifications