AuftragsverfolgungKontakt
Test kits, drop count tests | Avantor
Test kits, drop count tests
Ascorbic acid, model ASC-1, 10 - 200 mg/l, 1 KIT
Artikelnr. HACH2308100
Anbieter:   Hach
Test kits, drop count tests
Artikelnr. HACH2308100
Anbieter:  Hach
CAS Nummer:  
Test kits, drop count tests
Test kits, drop count tests
Ascorbic acid, model ASC-1, 10 - 200 mg/l, 1 KIT
CAS Nummer:  
Preis
183,00 €
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Achtung! Bitte beachten: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Spezifikationen
Spezifikationen
  • VE:
    1 KIT
  • Bezeichnung:
    Ascorbic acid, model ASC-1
  • Messbereich:
    10 - 200 mg/l
Spezifikationen