Propylenglycol ≥99%, TECHNICAL
Metalltrommel, 25 L
Anbieter: VWR Chemicals
Springe zu:
Product Details & Documents
Synonyme: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
- CAS Nummer:57-55-6
- Formel:CH₃CH(OH)CH₂OH
- MDL-Nummer:MFCD00064272
- Molekulargewicht:76.1 g/mol
- Merck-Index:13,07947
- Siedepunkt:187,6 °C (1013 hPa)
- Schmelzpunkt:–59 °C
- Dichte:1,04 g/cm³ (20 °C)
- Blitzpunkt:99 °C
- Lagertemperatur:Raumtemperatur
- EINECS:200-338-0
Documents
Zertifikate
Suche nach Chargennummer14515078.pdf 14515079.pdf P20_VWRC24414.296_SPEC_DE.pdf P20_VWRC24414.321_SPEC_DE.pdf P20_VWRC24414.365_SPEC_DE.pdf
Product specification
Mehr
1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
VWR Chemicals Big Packs and Bulks
Product Literature
Spezifikationen
Spezifikation Testergebnisse
- Assay:Min. 99 %
- Brechungsindex (20°C):1.431 - 1.435
Spezifikationen
- Packung:Metalltrommel
- VE:25 L
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



