Electrode Adapter Cables
Adapter, BNC meter input + screw cap electrode connector; detachable cable to BNC connector
Artikelnr. 662-1535
Anbieter: Thermo Orion
Springe zu:
Product Details & Documents
These cables can be used to convert an electrode with one connector to the required connector of another meter.
For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2 mm reference pins or just a stripped lead inputs.
Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors.
Spezifikationen
- Bezeichnung:Adapter, BNC meter input + screw cap electrode connector; detachable cable to BNC connector
- Seq:05