Express Eingabe
Austria
AuftragsverfolgungKontakt
Basins, Evaporating | Avantor

Basins, Evaporating

65 mm, 115 mm
Artikelnr. 216-2761
Anbieter:   SciLabware
Basins, Evaporating
Artikelnr. 216-2761
Anbieter:  SciLabware
Basins, Evaporating
Basins, Evaporating

65 mm, 115 mm

Preis
233,00 €
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents

PYREX®, borosilicate glass, with spout.

  • High resistance to chemical attack and thermal shock
  • Complies to DIN 12336
  • Quick and easy to clean

For wide range of applications. Feature flat base and curved sides.

Spezifikationen
  • Ext.Ø:
    115 mm
  • Höhe:
    65 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.