Abdampfschalen
73 ml, 19 mm, 70 mm
Artikelnr. BOCH8042
Anbieter: BOCHEM
Springe zu:
Product Details & Documents
Made of nickel 99,5 %.
- Good chemical resistance
- Very good temperatrure resistance
- Easy to clean
Without spout.
Spezifikationen
- Ext.Ø:70 mm
- Kapazität:73 ml
- Höhe:19 mm
- Seq:03