Mini molecular models: DNA
Mini molecular model of DNA, 22 layers
Artikelnr. 763-0454
Marketsource
Anbieter: SPIRING ENTERPRISES
Springe zu:
Product Details & Documents
Mini molecular models of DNA with nucleic acids, five-sided sugar molecules and pyramid-shaped phosphate molecules.
Only correct base pairs can be connected.
Ordering information: Components include colour-coded bases: G, C, A, T.
Spezifikationen
- Bezeichnung:Mini molecular model of DNA, 22 layers