Express Eingabe
Austria
AuftragsverfolgungKontakt
Heating tapes, TeMS2

Heating tapes, TeMS2

Heating tape TeMS6, 63 m length, 490 W
Catalog # ISOP184778-000
Supplier:   ISOPAD
Heating tapes, TeMS2
Catalog # ISOP184778-000
Supplier:  ISOPAD
Heating tapes, TeMS2
Heating tapes, TeMS2

Heating tape TeMS6, 63 m length, 490 W

Price
422,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Isopad IT-TeMS is a preterminated and PTFE insulated flexible heating tape.

  • Temperature resistant up to 260 °C
  • Non hazardous and used in ordinary areas
  • Parallel arrangement ensures a wide heat transfer surface

IT-TeMS2 is used in applications ranging from simple frost protection to temperature maintenance on short pipework and apparatus as well as for general container heating applications. TeMS4 or TeMS6 tapes can be used when more power is required.

Specifications
  • Bezeichnung:
    Heating tape TeMS6, 63 m length, 490 W
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.