γ-(N-Morpholino)propansulfonsäure (MOPS)
Glass bottle for solids, 100 g
Catalog # BMOL06170-1
Supplier: BIOMOL RESEARCH LABORATORIES
γ-(N-Morpholino)propansulfonsäure (MOPS)
Catalog # BMOL06170-1
Supplier: BIOMOL RESEARCH LABORATORIES
CAS Number: 1132-61-2
Jump to:
Product Details & Documents
Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
- CAS Number:1132-61-2
- Formula:C₇H₁₅NO₄S
- MDL Number:MFCD00006183
- Molecular Weight:209.27 g/mol
- Merck Index:13,06288
- Melting Pt:> 250 °C
- Flash Pt:110 °C (geschlossener Tiegel)
- Storage Temperature:Raumtemperatur
- EINECS:214-478-5
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



