Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Platinum wire

Platinum wire, Ø 0,6 mm
  631-7105
 :   CHEMPUR
Platinum wire
  631-7105
 :  CHEMPUR
Platinum wire

Platinum wire, Ø 0,6 mm

 
 
   
1 * 1 cm
 

Platinum 99,9%.

Platinum wire is used in many applications including gasket or O-ring fabrication, thermocouples, electrical contacts, loops and heater coils.

 
  • Bezeichnung:
    Platinum wire, Ø 0,6 mm
  • Seq:
    05