Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Platinum wire

Platinum wire, Ø 0,3 mm
  631-7102
 :   CHEMPUR
Platinum wire
  631-7102
 :  CHEMPUR
Platinum wire

Platinum wire, Ø 0,3 mm

 
250,00 €
   
1 * 50 cm
 

Platinum 99,9%.

Platinum wire is used in many applications including gasket or O-ring fabrication, thermocouples, electrical contacts, loops and heater coils.

 
  • Bezeichnung:
    Platinum wire, Ø 0,3 mm
  • Seq:
    02