Express Eingabe
Austria
AuftragsverfolgungKontakt
Connectors, PP | Avantor
Connectors, PP
Connector, straight, male luer slip/tubing nipple, 1,6 mm
Artikelnr. 229-0530
Anbieter:   PIEPER FILTER
Connectors, PP
Artikelnr. 229-0530
Anbieter:  PIEPER FILTER
Connectors, PP
Connectors, PP
Connector, straight, male luer slip/tubing nipple, 1,6 mm
Preis
36,00 €
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents
Connectors, made of PP, natural.
  • Good chemical resistance
  • Good temperature resistance
Spezifikationen
  • Schlauch Innen-Ø:
    1,6 mm
  • Bezeichnung:
    Connector, straight, male luer slip/tubing nipple
  • Seq:
    13
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.