Connectors, PP
Connector, straight, female luer lock/tubing nipple, 3,2 mm
229-0527
Connectors, made of PP, natural.
- Good chemical resistance
- Good temperature resistance
- Schlauch Innen-Ø:3,2 mm
- Bezeichnung:Connector, straight, female luer lock/tubing nipple
- Seq:09