Express Eingabe
Austria
AuftragsverfolgungKontakt
Connectors, PP
Connectors, PP
Connector, straight, female luer lock/tubing nipple, 1,6 mm
Catalog # 229-0525
Supplier:   PIEPER FILTER
Connectors, PP
Catalog # 229-0525
Supplier:  PIEPER FILTER
Connectors, PP
Connectors, PP
Connector, straight, female luer lock/tubing nipple, 1,6 mm
Price
22,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
Connectors, made of PP, natural.
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Schlauch Innen-Ø:
    1,6 mm
  • Bezeichnung:
    Connector, straight, female luer lock/tubing nipple
  • Seq:
    07
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.