Connectors, PP
Connector, straight, male luer lock/tubing nipple, 3,2 mm
Artikelnr. 229-0517
Anbieter: PIEPER FILTER
Springe zu:
Product Details & Documents
Connectors, made of PP, natural.
- Good chemical resistance
- Good temperature resistance
Spezifikationen
- Schlauch Innen-Ø:3,2 mm
- Bezeichnung:Connector, straight, male luer lock/tubing nipple
- Seq:03